missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Bottle Top Dispenser with Re-Circulation Valve

Provides a contemporary design coupled with a recirculation valve. Bottle Top Dispenser with Re-Circulation Valve has enhanced chemical compatibility and smooth piston movement.

Manufacturer:  Fisherbrand™ BEAT10

 View more versions of this product

Catalog No. 13681526



Description

Description

  • Re-circulation valve helps user in quick purging without wasting any reagent
  • Specially designed unique spring-less valve provides greater chemical compatibility, no chemical deposition and minimizing the maintainence
  • Presence of unique spring-less valve, PTFE piston and ETP O-ring provides greater chemical compatibility, ensures wiper like smooth and effortless piston movement
  • Specially designed 180° rotation volume setting knob provides quick and effortless volume adjustment
  • Adjustable delivery nozzle provides flexibility to users in demanding laboratory conditions
  • Bottle neck adapters of different sizes (28, 30, 32, 36, 40 and 45mm) ensures compatibility of dispenser with majority of reagent bottles
  • Each dispenser is accompanied with tool, which helps in easy and quick re-calibration
  • Contemporary and well designed contours, silky surface finish
  • Individually calibrated as per ISO 8655-2
Specifications

Specifications

1 to 10mL
  • CE Certificate
  • ISO 13485-2003
  • ISO 9001-2008
  • Compliance as per ISO 8655
  • Calibration lab certification by NABL as per ISO 17025
2.2g/cm3
Bottle Top Dispenser is used in laboratories to dispense reagent from the bottle in a preset volume accurately and safely into a receiving vessel.
Outer body is made up of PP (10% glass filled) which provides strength and high dimensional accuracy.
300mbar to 600mbar
Shape: Cylindrical
White and Fisher Blue
Bottle Top Dispenser
  • Calibration tool
  • Adapters set (5 nos.)
  • Instruction Manual
  • Inlet tube
  • Calibration report
500mm2
1 year free replacement warranty against any manufacturing defects.
Documents