missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Bottle Top Dispenser with Dual Inlet Technology
Dual Inlet Technology facilitates to re-fill the bottle and rinse the dispenser. Bottle Top Dispenser with Dual Inlet Technology provides greater chemical compatibility.
Description
- Dual Inlet Technology facilitates to re-fill the bottle, rinse the dispenser, perform dilution and dual reagent dispensing without dis-mounting the dispenser from its bottle
- Specially designed unique spring-less valve provides greater chemical compatibility, no chemical deposition and no maintenence required
- Presence of a unique spring-less valve, PTFE piston and ETP O-ring provides greater chemical compatibility, also ensures wiper like smooth and effortless piston movement
- Specially designed 180° rotation volume setting knob provides quick and effortless volume adjustment
- Adjustable delivery nozzle provides flexibility to the users in demanding laboratory conditions
- Bottle neck adapters of different sizes (28, 30, 32, 36, 40 and 45mm) ensures compatibility of dispenser with majority of reagent bottles
- Each dispenser is accompanied with tool, which helps in easy and quick re-calibration Contemporary and well designed contours, silky surface finish
- Individually calibrated as per ISO 8655-2
Specifications
Bottletop Dispenser | |
CE Certificate, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655, Calibration lab certification by NABL as per ISO 17025 | |
2.2 g/cm3 | |
Polypropylene - Outer Body (10% Glass Filled) | |
Bottle Top Dispenser is used in laboratories to dispense reagent from the bottle in a preset volume accurately and safely into a receiving vessel. | |
300mbar to 600mbar | |
1 year free replacement warranty against any manufacturing defects. |
Only the piston needs to be removed for autoclaving the instrument. Piston is also autoclavable | |
White and Fisher Blue | |
Shape: Cylindrical | |
5 to 60 mL | |
Calibration tool, Adapters set (4 nos.), Instruction Manual 4, Inlet tube, Calibration report A, Calibration report B, Inlet tube for the second inlet, Caps for the second bottle (4 nos.) | |
500mm2 |