Please login to your online account to display your discounted pricing

Bosutinib, Tocris™

Dual Src-Abl inhibitor; antiproliferative

$310.33 - $1,282.40

Chemical Identifiers

Chemical Name or Material Bosutinib
Assay Percent Range >99%
Molecular Formula C26H29Cl2N5O3
Formula Weight 530.45
Synonym 4-[(2, 4-Dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methyl-1-piperazinyl)propoxy]-3-quinolinecarbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

436110

 
r & d systems
436110
10mg Each for $310.33

436150

 
r & d systems
436150
50mg Each for $1,282.40
Description & Specifications

Specifications

Formula Weight 530.45
Quantity 10mg
CAS 380843-75-4
Recommended Storage Store at +4°C
Assay Percent Range >99%
Synonym 4-[(2, 4-Dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methyl-1-piperazinyl)propoxy]-3-quinolinecarbonitrile
Chemical Name or Material Bosutinib
Molecular Formula C26H29Cl2N5O3
Safety and Handling

Store at +4°C