Fisherbrand™ Custom Cleaned Clear Boston Round Bottles
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Custom Cleaned Clear Boston Round Bottles are manufactured from clear soda glass.
$148.83 - $423.83
Specifications
Cleaning Procedure USP purified Water
Closure Color White
Closure Included Yes
Closure Liner Polyethylene
Closure Material PTFE
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-912-009
View Documents Promotion Details
Fisherbrand™
02912009
30 mL
Case of 48 Each for $406.69
02-912-010
View Documents Promotion Details
Fisherbrand™
02912010
60 mL
Case of 24 Each for $423.83
02-912-012
View Documents Promotion Details
Fisherbrand™
02912012
250 mL
Case of 12 Each for $148.83
02-912-013
View Documents Promotion Details
Fisherbrand™
02912013
500 mL
Case of 12 Each for $213.00
02-912-014
View Documents Promotion Details
Fisherbrand™
02912014
1000 mL
Case of 12 Each for $251.28
Description
Description
  • Narrow mouth
  • Pre-cleaned for visible particulates using deionised water
  • With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications
Specifications
USP purified Water
Yes
PTFE
Narrow
Clear
Boston Round Bottle
White
Polyethylene
Screw Cap
Round
Soda Lime Glass