Sartorius™ Entris™ 90 mm Analytical Balances
Provides speedy results for heavy workloads with repetitive applications. Sartorius™ Entris™ 90mm Analytical Balances feature a new operating concept: easy configuration of individual settings guided by short, menu-driven text prompts and simple navigation using cursor keys.
Manufacturer:  Sartorius™ ENTRIS64I1SUS
 View more versions of this product
Catalog No. 14-559-016
Description
Description
  • White-backlit, high-contrast display with 15mm digits; exceptionally easy to read under any room lighting conditions
  • Choice of 5 languages
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Specifications
Specifications
Internal
90mm
0.1 mg
176 x 173 x 240mm
0.2mg
0.1mg
60g
Analytical Balance
2.5 sec.
330 x 230 x 330mm
ENTRIS64i-1S