Please login to your online account to display your discounted pricing

Boron trifluoride tetrahydrofuran complex 50%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC388631000

 View more versions of this product

Catalog No. AC388631000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 139.91
Physical Form Viscous Liquid
Quantity 100mL
Color Brown
Assay 48 to 50%
Density 1.2700g/mL
Identification Confirmed
Chemical Name or Material Boron trifluoride tetrahydrofuran complex
Packaging Glass bottle
CAS 462-34-0
Molecular Formula BF3·C4H8O
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Assay Percent Range 50%
Health Hazard 1 Danger
MDL Number MFCD00040372
Health Hazard 2 GHS P Statement Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Harmful if inhaled.

Specifications

Chemical Name or Material Boron trifluoride tetrahydrofuran complex
CAS 462-34-0
Assay 48 to 50%
Assay Percent Range 50%
Molecular Formula BF3·C4H8O
Formula Weight 139.91
MDL Number MFCD00040372
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS P Statement Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Harmful if inhaled.

SDS & Safety Documents