Please login to your online account to display your discounted pricing

Boron trifluoride tetrahydrofuran complex 50%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Boron trifluoride tetrahydrofuran complex
CAS 462-34-0
Assay 48 to 50%
Assay Percent Range 50%
Molecular Formula BF3·C4H8O
Formula Weight 139.91
MDL Number MFCD00040372
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388635000

  SDS
thermo fisher scientific
AC388635000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC388631000

  SDS
thermo fisher scientific
AC388631000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid
Formula Weight 139.91
Quantity 500mL
Color Brown
Assay 48 to 50%
Density 1.2700g/mL
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Assay Percent Range 50%
Molecular Formula BF3·C4H8O
CAS 462-34-0
Packaging Glass bottle
Identification Confirmed
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Harmful if inhaled.
Chemical Name or Material Boron trifluoride tetrahydrofuran complex
MDL Number MFCD00040372

Lewis acid

Safety and Handling

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS P Statement Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Harmful if inhaled.