Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron trifluoride-ethylamine complex, 95%

Chemical Identifiers

Chemical Name or Material Boron trifluoride-ethylamine complex
CAS 75-23-0
Molecular Formula C2H7BF3N
Linear Formula BF3·NH2CH2CH3
Formula Weight 112.89g/mol
MDL Number MFCD00144277
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1434022

 
alfa aesar™
L14340-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point ≈85°C
Quantity 100g
Flash Point 157°C (314°F)
Odor Fish-like
Chemical Name or Material Boron trifluoride-ethylamine complex
CAS 75-23-0
Assay 95%
Molecular Formula C2H7BF3N
Linear Formula BF3·NH2CH2CH3
Formula Weight 112.89g/mol
MDL Number MFCD00144277
UN Number UN2811
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number KQ8060000
TSCA TSCA

Applications
Boron trifluoride-ethylamine complex is used as latent curing agent for epoxy resin. It is also used as pharmaceutical intermediate and in chemical research.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.