Learn More
Boron trifluoride etherate, approx. 48% BF3, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 174560025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Boron trifluoride etherate | |
109-63-7 | |
Colorless to Straw-Yellow | |
126.0°C | |
Glass bottle | |
1.3429 to 1.3449 | |
MFCD00013194 | |
01,70; 02,35; 03,33; 04,44; 05,52; 06,65; 07,31; 08,51; 09,64; 10,52; 11,72; 12,66; 13,3; 14,54; 15,45; 16,44; 17,52 | |
15,1352 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in ether | |
FB(F)F.CCOCC | |
141.93 | |
1.89 mPa.s (20°C) | |
approx. 48% BF3 |
48% BF3 | |
-60.0°C | |
1.1200g/mL | |
58°C | |
C4H10BF3O | |
(C2H5)2O·BF3 | |
2.5 kg | |
1.12 | |
Boron trifluoride ethyl ether | |
KZMGYPLQYOPHEL-UHFFFAOYSA-N | |
ethoxyethane;trifluoroborane | |
8000 | |
141.93 | |
Liquid, Fuming In Moist Air |
Chemical Identifiers
109-63-7 | |
141.93 | |
KZMGYPLQYOPHEL-UHFFFAOYSA-N | |
8000 | |
FB(F)F.CCOCC |
C4H10BF3O | |
MFCD00013194 | |
Boron trifluoride ethyl ether | |
ethoxyethane;trifluoroborane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 203-689-8
TSCA : TSCA
Recommended Storage : Product darkens on storage
RUO – Research Use Only