missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron trifluoride dimethyl etherate, Acros Organics™

Manufacturer:  Acros Organics AC204062500

 View more versions of this product

Catalog No. AC204062500



Description

Description

Lewis acid
Specifications

Specifications

Boron trifluoride dimethyl etherate
62°C
Glass bottle
1.23
Brown
250mL
C2H6BF3O
MFCD00013192
FOWZWMGLMYVYDH-UHFFFAOYSA-N
dimethyloxonio(trifluoro)boranuide
2724165
Liquid, Fuming In Moist Air
1.2300g/mL
Authentic
1.3070 to 1.3090
126.0°C to 127.0°C
-14.0°C
353-42-4
(CH3)2O·BF3
boron trifluoride dimethyl ether complex
[B-]([O+](C)C)(F)(F)F
113.874
113.87
≈60% (BF3)

Chemical Identifiers

353-42-4
113.874
FOWZWMGLMYVYDH-UHFFFAOYSA-N
2724165
[B-]([O+](C)C)(F)(F)F
C2H6BF3O
MFCD00013192
boron trifluoride dimethyl ether complex
dimethyloxonio(trifluoro)boranuide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-532-1

RTECSNumber : ED8400000

TSCA : TSCA

SDS Documents