missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron trifluoride dimethyl etherate, ACROS Organics™

$278.79 - $278.79

Chemical Identifiers

CAS 353-42-4
Molecular Formula C2H6BF3O
Molecular Weight (g/mol) 113.874
MDL Number MFCD00013192
InChI Key FOWZWMGLMYVYDH-UHFFFAOYSA-N
Synonym boron trifluoride dimethyl ether complex, boron trifluoride dimethyl etherate, boron trifluoride methyl etherate, dimethyl ether trifluoroboron, dimethyloxo trifluoro-??-borane, dimethyloxonio trifluoro boranuide
PubChem CID 2724165
IUPAC Name dimethyloxonio(trifluoro)boranuide
SMILES [B-]([O+](C)C)(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC204062500
SDS View Documents
Thermo Fisher Scientific
AC204062500
250mL Glass bottle
Each for $278.79
 
Description

Description

Lewis acid
Specifications

Chemical Identifiers

353-42-4
113.874
FOWZWMGLMYVYDH-UHFFFAOYSA-N
2724165
[B-]([O+](C)C)(F)(F)F
C2H6BF3O
MFCD00013192
boron trifluoride dimethyl ether complex, boron trifluoride dimethyl etherate, boron trifluoride methyl etherate, dimethyl ether trifluoroboron, dimethyloxo trifluoro-??-borane, dimethyloxonio trifluoro boranuide
dimethyloxonio(trifluoro)boranuide

Specifications

1.2300g/mL
Authentic
1.3070 to 1.3090
126.0°C to 127.0°C
-14.0°C
353-42-4
(CH3)2O·BF3
boron trifluoride dimethyl ether complex, boron trifluoride dimethyl etherate, boron trifluoride methyl etherate, dimethyl ether trifluoroboron, dimethyloxo trifluoro-??-borane, dimethyloxonio trifluoro boranuide
[B-]([O+](C)C)(F)(F)F
113.874
113.87
≈60% (BF3)
62°C
Glass bottle
1.23
Brown
250mL
C2H6BF3O
MFCD00013192
FOWZWMGLMYVYDH-UHFFFAOYSA-N
dimethyloxonio(trifluoro)boranuide
2724165
Liquid, Fuming In Moist Air
Boron trifluoride dimethyl etherate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-532-1

RTECSNumber : ED8400000

TSCA : TSCA

SDS