Please login to your online account to display your discounted pricing

Boron trifluoride dimethanol complex, 50-52 wt% BF3, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  158900050

 View more versions of this product

Catalog No. AC158900050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥50% (BF3)
Formula Weight 131.89
Quantity 5g
Color Clear colorless to slightly yellow
EINECS Number 220-543-9
TSCA TSCA
Chemical Name or Material Boron trifluoride dimethanol complex
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
CAS 7637-07-2
Name Note 50-52 wt% BF3
Molecular Formula C2H8BF3O2
Linear Formula BF3·2CH3OH
MDL Number MFCD00071635
CAS 2802-68-8
Density (20°C) 1.21 to 1.235kg/L
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 67-56-1

Specifications

Chemical Name or Material Boron trifluoride dimethanol complex
Name Note 50-52 wt% BF3
CAS 2802-68-8
CAS 7637-07-2
CAS 67-56-1
Assay ≥50% (BF3)
Molecular Formula C2H8BF3O2
Linear Formula BF3·2CH3OH
Formula Weight 131.89
MDL Number MFCD00071635
Accessories

Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.

SDS & Safety Documents