missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron trifluoride dimethanol complex, 50-52 wt% BF3, ACROS Organics™

Manufacturer:  Acros Organics 158900010

 View more versions of this product

Catalog No. AC158900010



Description

Description

Lewis acid
Specifications

Specifications

Boron trifluoride dimethanol complex
68°C
1.34
59.0°C (4.0 mmHg)
-20.0°C
50-52 wt% BF3
CH4BF3O
MFCD00071635
Solubility in water: hydrolysis.
B(F)(F)F.CO
99.847
131.89
≥50% (BF3)
1.2220g/mL
Glass bottle
1.222
Colorless to Yellow
1kg
2802-68-8,7637-07-2,67-56-1
BF3·2CH3OH
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
JBXYCUKPDAAYAS-UHFFFAOYSA-N
methanol;trifluoroborane
11062313
Liquid

Chemical Identifiers

2802-68-8
99.847
JBXYCUKPDAAYAS-UHFFFAOYSA-N
11062313
B(F)(F)F.CO
CH4BF3O
MFCD00071635
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
methanol;trifluoroborane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 220-543-9

TSCA : TSCA

SDS Documents