Learn More
Boron trifluoride-dimethanol complex, 50-52% w/w boron trifluoride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B21357AK
Description
It finds it application as a reagent for esterification of acids and also for the selective cleavage of trityl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds it application as a reagent for esterification of acids and also for the selective cleavage of trityl ethers.
Solubility
Hydrolyzes in water.
Notes
^=50% boron fluoride in methanol
Specifications
Boron trifluoride-dimethanol complex | |
1.222 | |
68°C (154°F) | |
MFCD00071635 | |
UN2922 | |
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution | |
MPRMFRXCVMCOMX-UHFFFAOYSA-N | |
boron(3+) hydrogen methanol methanolate trifluoride | |
11062313 | |
50 to 52% w/w boron trifluoride |
2802-68-8 | |
58°C to 60°C (4 mmHg) | |
C2H8BF3O2 | |
250 mL | |
Moisture sensitive | |
Hydrolyzes in water. | |
CO.CO.FB(F)F | |
131.89 | |
131.89 |
Chemical Identifiers
2802-68-8 | |
131.89 | |
MPRMFRXCVMCOMX-UHFFFAOYSA-N | |
11062313 |
C2H8BF3O2 | |
MFCD00071635 | |
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution | |
CO.CO.FB(F)F |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H311+H331-H314-H335-H370-H372
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 220-543-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only