missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron trifluoride diethyl etherate, 98+%

Manufacturer:  Alfa Aesar™ A15275AU

 View more versions of this product

Catalog No. AAA15275AU



Description

Description

Applications
Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

Solubility
Miscible with ether and alcohol.

Notes
Moisture sensitive. Store in cool place. Recommended to use under inet atomosphere. Incompatible with metals, acids, bases, alcohols, alkali metals, oxidizing agents and water. Reacts violently with water.
Specifications

Specifications

Boron trifluoride diethyl etherate
Acrid
-60°C
109-63-7
BF3€O(CH2CH3)2
UN2604
14,1350
KZMGYPLQYOPHEL-UHFFFAOYSA-N
ethoxyethane;trifluoroborane
8000
≥98%
47°C (116°F)
125°C to 126°C
1000mL
C4H10BF3O
MFCD00013194
3909607
Boron trifluoride etherate; Boron fluoride-ether
B(F)(F)F.CCOCC
141.928
141.93

Chemical Identifiers

109-63-7
141.928
KZMGYPLQYOPHEL-UHFFFAOYSA-N
8000
B(F)(F)F.CCOCC
C4H10BF3O
MFCD00013194
Boron trifluoride etherate; Boron fluoride-ether
ethoxyethane;trifluoroborane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H372-H314-H318-H226-H302
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIFLUORIDE DIETHYL ETHERATE

TSCA : TSCA

Documents