Please login to your online account to display your discounted pricing

Boron trifluoride - acetic acid complex, ca. 33% BF3, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC277220025

 View more versions of this product

Catalog No. AC277220025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 187.92
Color Clear light yellow to yellow-brown
Physical Form Liquid
Quantity 2.5L
TSCA TSCA
EINECS Number 206-768-5
Assay 33 to 37% (BF3)
Density 1.345g/mL
Fieser 16,176
Beilstein 04, 115
Storage Information Product darkens on standing
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Free Acid 63 to 67%
Health Hazard 1 GHS Signal Word: Danger
Linear Formula BF3·CH3COOH
Name Note ca. 33% BF3
Molecular Formula BF3·C4H8O4
Chemical Name or Material Boron trifluoride - acetic acid complex
CAS 373-61-5
MDL Number MFCD00036359
Packaging Glass bottle
Synonym Trifluoroborane compound with acetic acid (1:2)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Boron trifluoride - acetic acid complex
Name Note ca. 33% BF3
CAS 373-61-5
Assay 33 to 37% (BF3)
Molecular Formula BF3·C4H8O4
Linear Formula BF3·CH3COOH
Formula Weight 187.92
MDL Number MFCD00036359
Synonym Trifluoroborane compound with acetic acid (1:2)
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

SDS & Safety Documents