missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron trifluoride, 99+%

Manufacturer:  Alfa Aesar™ 2287422

 View more versions of this product

Catalog No. AA2287422



Specifications

Specifications

Boron trifluoride
BF3
UN1008
boron trifluoride, borane, trifluoro, boron fluoride, trifluoroboron, boron fluoride, fluorure de bore, fluorure de bore french, unii-7jgd48px8p, 7jgd48px8p, trifluoro boron
B(F)(F)F
67.81
CHEBI:33093
Gas
3.076g/mL
Moisture Sensitive
Colorless
100g
7637-07-2
MFCD00011316
14,1349
WTEOIRVLGSZEPR-UHFFFAOYSA-N
trifluoroborane
6356
67.81
≥99%
Pungent
-100°C
-127°C

Chemical Identifiers

7637-07-2
67.81
WTEOIRVLGSZEPR-UHFFFAOYSA-N
6356
trifluoroborane
BF3
MFCD00011316
boron trifluoride, borane, trifluoro, boron fluoride, trifluoroboron, boron fluoride, fluorure de bore, fluorure de bore french, unii-7jgd48px8p, 7jgd48px8p, trifluoro boron
CHEBI:33093
B(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310-P302+P352-P403
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 2.3; Proper Shipping Name: BORON TRIFLUORIDE

RTECSNumber : ED2275000

TSCA : TSCA

SDS Documents