missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron trifluoride, 12% (1.5M) in methanol, ACROS Organics™

Boron trifluoride, 12% (1.5M) in methanol, ACROS Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3)

Manufacturer:  Thermo Fisher Scientific AC402760010

 View more versions of this product

Catalog No. AC402760010



Description

Description

Boron trifluoride, 12% (1.5M) in methanol, ACROS Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3)

Acylation reagent, Lewis acid
Specifications

Specifications

Boron trifluoride
373-57-9,67-56-1,373-57-9
BF3
14, 1349
Solubility in water: may decompose
B(F)(F)F.CO
67.81
67.81
10 to 15% (w/v BF3)
Glass bottle
0.87
Amber to Colorless
1kg
1.5M solution in methanol
BF3
MFCD00071635
.bf3 in methanol, bf3 meoh, bf3 methanol, boron fluoride methanol, boron trifluoride methanol, boron trifluoride solution, boron trifluoride-methanol complex, boron trifluoride-methanol solution, boron trifluoride-methanol solution in methanol, borontrifluoride methanol
JBXYCUKPDAAYAS-UHFFFAOYSA-N
methanol;trifluoroborane
11062313
Solution
0.8700g/mL
Solubility in water: may decompose.
65.0°C
-98.0°C
4°C

Chemical Identifiers

373-57-9
67.81
JBXYCUKPDAAYAS-UHFFFAOYSA-N
11062313
B(F)(F)F.CO
BF3
MFCD00071635
.bf3 in methanol, bf3 meoh, bf3 methanol, boron fluoride methanol, boron trifluoride methanol, boron trifluoride solution, boron trifluoride-methanol complex, boron trifluoride-methanol solution, boron trifluoride-methanol solution in methanol, borontrifluoride methanol
methanol;trifluoroborane
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Toxic if swallowed.
Reacts violently with water.
Toxic in contact with skin.
Causes damage to organs

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 206-766-4

RTECSNumber : ED2275000

TSCA : TSCA

SDS Documents