Please login to your online account to display your discounted pricing

Boron trifluoride, 12% (1.5M) in methanol, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  402761000

 View more versions of this product

Catalog No. AC402761000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 10 to 15% (w/v BF3)
Physical Form Solution
Formula Weight 67.81
Quantity 100g
Color Colorless to amber
Merck Index 14, 1349
EINECS Number 231-569-5
TSCA TSCA
RTECS Number ED2275000
Density 0.8700g/mL
Chemical Name or Material Boron trifluoride
Name Note 1.5M solution in methanol
Molecular Formula BF3
Linear Formula BF3
MDL Number MFCD00071635
CAS 373-57-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic if swallowed.
Reacts violently with water.
CAS 67-56-1
CAS 373-57-9
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Boron trifluoride
Name Note 1.5M solution in methanol
CAS 373-57-9
CAS 67-56-1
CAS 373-57-9
Assay 10 to 15% (w/v BF3)
Molecular Formula BF3
Linear Formula BF3
Formula Weight 67.81
MDL Number MFCD00071635
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic if swallowed.
Reacts violently with water.

SDS & Safety Documents