missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 176681000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Boron trichloride | |
1M Solution in Methylene Chloride | |
BCl3 | |
MFCD00011313 | |
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44 | |
15, 1350 | |
Solubility in water: may decompose. | |
ClB(Cl)Cl | |
117.16 | |
117.17 | |
1.326 | |
Liquid |
Colorless to Brown | |
10294-34-5,75-09-2 | |
BCl3 | |
0.9 to 1.1M | |
1.3260g/mL | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
trichloroborane | |
25135 | |
AcroSeal™ Glass bottle | |
100 mL |
RUO – Research Use Only