Learn More
Boron trichloride, 1M soln. in hexanes, mixed isomers, Thermo Scientific Chemicals
$141.42 - $429.56
Chemical Identifiers
CAS | 10294-34-5 |
---|---|
Molecular Formula | BCl3 |
Molecular Weight (g/mol) | 117.16 |
MDL Number | MFCD00011313 |
InChI Key | FAQYAMRNWDIXMY-UHFFFAOYSA-N |
Synonym | boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas |
PubChem CID | 25135 |
IUPAC Name | trichloroborane |
SMILES | ClB(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH32833AE
|
Thermo Scientific Chemicals
H32833AE |
100 mL |
Each for $141.42
|
|
|||||
AAH32833AP
|
Thermo Scientific Chemicals
H32833AP |
500 mL |
Each for $429.56
|
|
|||||
Description
A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat. Unstable. Store at 2-8°C.
Chemical Identifiers
10294-34-5 | |
117.16 | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
25135 | |
ClB(Cl)Cl |
BCl3 | |
MFCD00011313 | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
trichloroborane |
Specifications
10294-34-5 | |
BCl3 | |
MFCD00011313 | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
trichloroborane | |
25135 | |
1M soln. in hexanes | |
0.738 |
100 mL | |
BCl3 | |
UN3286 | |
Reacts with water. | |
ClB(Cl)Cl | |
117.16 | |
117.17 | |
Moisture sensitive; May form a precipitate | |
Boron trichloride, mixed isomers |
Safety and Handling
GHS H Statement
H225-H300-H331-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Fatal if swallowed.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P331-P363-P372-P374-P380-P501c
H225-H300+H330-H314-H318-H361-H373-X
DOTInformation : Hazard Class: 3; Packaging Group: I
EINECSNumber : 233-658-4
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only