missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron tribromide dimethylesulfide, 1M solution in methylene chloride, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC428328000

 View more versions of this product

Catalog No. AC428328000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Boron tribromide dimethylesulfide
29957-59-3,75-09-2
312.66
Dimethylesulfide-tribromoborane
AcroSeal™ Glass Bottle
1M solution in methylene chloride
C2H6BBr3S
MFCD00043296
1.456g/mL
800mL

Chemical Identifiers

29957-59-3,75-09-2
312.66
Dimethylesulfide-tribromoborane
C2H6BBr3S
MFCD00043296
Boron tribromide dimethylesulfide, 1M solution in methylene chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Suspected of causing cancer.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger