Please login to your online account to display your discounted pricing

Boron tribromide dimethylesulfide, 1M solution in methylene chloride, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC428321000

 View more versions of this product

Catalog No. AC428321000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 312.66
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 1.456g/mL
MDL Number MFCD00043296
Synonym Dimethylesulfide-tribromoborane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Boron tribromide dimethylesulfide
Molecular Formula C2H6BBr3S
CAS 29957-59-3
CAS 75-09-2
Name Note 1M solution in methylene chloride
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.

Specifications

Chemical Name or Material Boron tribromide dimethylesulfide
Name Note 1M solution in methylene chloride
CAS 29957-59-3
CAS 75-09-2
Molecular Formula C2H6BBr3S
Formula Weight 312.66
MDL Number MFCD00043296
Synonym Dimethylesulfide-tribromoborane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.

SDS & Safety Documents