Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron tribromide, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Boron tribromide
CAS 10294-33-4
Formula Weight 250.52
MDL Number MFCD00011312
UN Number UN2692
Merck Index 14,1347
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3584109

  SDS
alfa aesar™
3584109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3584118

  SDS
alfa aesar™
3584118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.52
CAS 10294-33-4
Density 2.643g/mL
Boiling Point 90°C
Health Hazard 1 Danger
Physical Form Liquid
TSCA TSCA
Assay Percent Notes (metals basis)
Melting Point −46°C
Health Hazard 2 GHS H Statement
H300-H330-H314-H318
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Odor Acrid
UN Number UN2692
Merck Index 14,1347
DOT Information Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIBROMIDE
MDL Number MFCD00011312
Assay 99.999%
Sensitivity Moisture Sensitive
RTECS Number ED7400000
Quantity 10g
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Light yellow
Chemical Name or Material Boron tribromide
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H314-H318
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.