missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron tribromide, 99.9%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 295201000
Description
C-X bond formation reagent, Lewis acidSpecifications
Boron tribromide | |
free bromine (Br2): < 0.13% | |
BBr3 | |
MFCD00011312 | |
15, 1349 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol | |
B(Br)(Br)Br | |
250.52 | |
250.52 | |
99.9% | |
100ppm max. | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol | |
100ppm max. | |
Amber | |
100g |
10294-33-4 | |
99.85% min. (By difference) | |
BBr3 | |
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43 | |
borane, tribromo, boron bromide, boron tri bromide, boron tri-bromide, boron tribromide, boron tribrornide, hsdb 327, tribromoboran, tribromoboron | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
tribromoborane | |
25134 | |
Liquid | |
2.6500g/mL | |
Glass bottle | |
2.65 | |
91.2°C | |
-46.0°C |
Chemical Identifiers
10294-33-4 | |
250.52 | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
25134 | |
B(Br)(Br)Br |
BBr3 | |
MFCD00011312 | |
borane, tribromo, boron bromide, boron tri bromide, boron tri-bromide, boron tribromide, boron tribrornide, hsdb 327, tribromoboran, tribromoboron | |
tribromoborane |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 233-657-9