Please login to your online account to display your discounted pricing

Boron tribromide, 1M solution in methylene chloride, ACROS Organics™


Quantity 800mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC198908000

 View more versions of this product

Catalog No. AC198908000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 250.52
Quantity 800mL
Color Clear yellow to dark brown
Concentration 0.9 to 1.1M
Density 1.4670g/mL
Fieser 01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43
Merck Index 15, 1349
Linear Formula BBr3
CAS 75-09-2
Chemical Name or Material Boron tribromide
Molecular Formula BBr3
MDL Number MFCD00011312
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
Suspected of causing cancer.
Reacts violently with water.
Packaging Glass bottle
Storage Information Darkens on storage
Name Note 1M Solution in Methylene Chloride
CAS 10294-33-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Boron tribromide
Name Note 1M Solution in Methylene Chloride
CAS 10294-33-4
CAS 75-09-2
Molecular Formula BBr3
Linear Formula BBr3
Formula Weight 250.52
MDL Number MFCD00011312
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
Suspected of causing cancer.
Reacts violently with water.

SDS & Safety Documents