missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron tribromide, 1M solution in methylene chloride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 198900100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acidSpecifications
Boron tribromide | |
10294-33-4,75-09-2 | |
Solution | |
BBr3 | |
MFCD00011312 | |
15, 1349 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in alcohol and ccl4 | |
BrB(Br)Br | |
250.52 | |
250.52 | |
Glass bottle | |
1.4670g/mL |
1M Solution in Methylene Chloride | |
Yellow to Brown | |
10 mL | |
BBr3 | |
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43 | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
tribromoborane | |
25134 | |
0.9 to 1.1M | |
1.467 |
RUO – Research Use Only