missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron tribromide, 1M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 378191000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
90.0°C | |
10294-33-4,142-82-5 | |
BBr3 | |
−1°C | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
tribromoborane | |
25134 | |
AcroSeal™ Glass bottle | |
0.8690g/mL |
Boron tribromide | |
100 mL | |
MFCD00011312 | |
15,1349 | |
Solubility in water: reacts | |
BrB(Br)Br | |
250.52 | |
250.52 | |
0.869 |
RUO – Research Use Only