Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron silicide, 98% (metals basis)

Chemical Identifiers

Chemical Name or Material Boron silicide
CAS 12008-29-6
Formula Weight 92.95
MDL Number MFCD00180907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3567009

  SDS
alfa aesar™
3567009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3567018

  SDS
alfa aesar™
3567018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 92.95
Odor Odorless
Mesh Size −200 Mesh
Chemical Name or Material Boron silicide
Physical Form Powder
Assay 98%
TSCA TSCA
MDL Number MFCD00180907
Assay Percent Notes (metals basis)
CAS 12008-29-6
Quantity 10g