Please login to your online account to display your discounted pricing

Boron, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg B) Solute: Boric Acid Solvent: Distilled Water

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSB155500

 View more versions of this product

Catalog No. SB155-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Melting Point 0°C
Quantity 500mL
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Viscosity [NULL]
Density 1.0g/cm³
Chemical Name or Material Boron, Reference Standard Solution
Name Note 1000ppm ±1%
Concentration 990 to 1010ppm
MDL Number MFCD00134034
CAS 10043-35-3
ChemAlert Storage Symbol Gray
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
CAS 7732-18-5
Packaging Poly Bottle

Specifications

Chemical Name or Material Boron, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 10043-35-3
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134034
Accessories
 

SDS & Safety Documents