Please login to your online account to display your discounted pricing

Boron, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg B) Solute: Boric Acid Solvent: Distilled Water

$93.65 - $116.00

Chemical Identifiers

Chemical Name or Material Boron, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 10043-35-3
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134034
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB155-100

  SDS
fisher scientific company
FLSB155100
Poly Bottle 100mL Each for $116.00
Case of 6 Each for $696.00

SB155-500

  SDS
fisher scientific company
FLSB155500
Poly Bottle 500mL Each for $93.65
Case of 6 Each for $561.90
Description & Specifications

Specifications

Melting Point 0°C
Purity Grade Certified Grade
Quantity 100mL
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Color Colorless
Concentration 990 to 1010ppm
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
MDL Number MFCD00134034
ChemAlert Storage Symbol Gray
CAS 7732-18-5
Packaging Poly Bottle
CAS 10043-35-3
Chemical Name or Material Boron, Reference Standard Solution
Name Note 1000ppm ±1%
Safety and Handling