missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron powder, crystalline, -4+40 mesh, Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 1011206

 View more versions of this product

Catalog No. AA1011206



Specifications

Specifications

Boron powder
7440-42-8
B
UN3178
and compounds, boracium, boranylidyne, boro, boron, amorphous, boron, elemental, boron, metallic, boron-elemental, powder, crystalline, unii-n9e3x5056q
[B]
Odorless
5462311
Crystalline Powder
Puratronic™
5 g
(metals basis)
MFCD00134034
−4+40 Mesh
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron
10.81
CHEBI:27560
99.9999%

Chemical Identifiers

7440-42-8
10.81
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
5462311
boron
B
MFCD00134034
and compounds, boracium, boranylidyne, boro, boron, amorphous, boron, elemental, boron, metallic, boron-elemental, powder, crystalline, unii-n9e3x5056q
CHEBI:27560
[B]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H332-H335
Flammable solid.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-151-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents