Learn More
Alfa Aesar™ Boron powder, crystalline, -4+40 mesh, 99.999% (metals basis)
Manufacturer: Alfa Aesar™ 1282003
Specifications
Boron powder | |
Brown to Black | |
7440-42-8 | |
(metals basis) | |
MFCD00134034 | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
[B] | |
10.81 | |
CHEBI:27560 | |
99.999% |
−4+40 Mesh | |
1g | |
Odorless | |
B | |
UN3178 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
boron | |
5462311 | |
Crystalline Powder |
Chemical Identifiers
7440-42-8 | |
10.81 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
5462311 | |
boron |
B | |
MFCD00134034 | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
CHEBI:27560 | |
[B] |
Safety and Handling
GHS H Statement
H228-H302-H332-H335
Flammable solid.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
RTECSNumber : ED7350000
TSCA : TSCA