missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron powder (96+ atomic % B-11), crystalline, -100 mesh, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4425706

 View more versions of this product

Catalog No. AA4425706



Specifications

Specifications

Boron powder
7440-42-8
B
UN3178
and compounds
[B]
Odorless
5462311
Powder
5 g
(metals basis), (≥96 atomic % B-11)
MFCD00134034
−100 Mesh
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron
10.81
CHEBI:27560
99.9%

Chemical Identifiers

7440-42-8
10.81
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
5462311
boron
B
MFCD00134034
and compounds
CHEBI:27560
[B]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H332-H335
Flammable solid.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-151-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents