Please login to your online account to display your discounted pricing

Boron powder, 94-96%, Alfa Aesar™

$158.22 - $638.86

Chemical Identifiers

Chemical Name or Material Boron powder
Name Note For analysis APS >5 micron
CAS 7440-42-8
Physical Form Powder
Assay Percent Range 94 to 96%
MDL Number MFCD00134034
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3968018

  SDS
alfa aesar
3968018
50g Each for $158.22

AA3968030

  SDS
alfa aesar
3968030
250g Each for $638.86
Description & Specifications

Specifications

Quantity 50g
EINECS Number 231-151-2
Physical Form Powder
Chemical Name or Material Boron powder
MDL Number MFCD00134034
CAS 7440-42-8
Name Note For analysis APS >5 micron
Assay Percent Range 94 to 96%