missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron pieces, crystalline, 1-30mm (0.039-1.2in), 99.4% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04185830
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boron | |
7440-42-8 | |
250 g | |
B | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
[B] | |
10.81 | |
CHEBI:27560 | |
Odorless |
Gray | |
Crystalline Pieces | |
99.4% (Metals basis) | |
MFCD00134034 MFCD00151272 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
boron | |
5462311 | |
700°C (1292°F) | |
Dark Gray pieces |
Chemical Identifiers
7440-42-8 | |
10.81 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
5462311 | |
boron |
B | |
MFCD00134034 MFCD00151272 | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
CHEBI:27560 | |
[B] |
Safety and Handling
H302; Harmful if swallowed.
EINECSNumber : 231-151-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only