Learn More
Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04178618
Description
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants and dopants in semiconductor materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1860°C | |
∽450°C | |
White | |
50 g | |
B2O3 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
oxo[(oxoboranyl)oxy]borane | |
518682 | |
69.62 | |
Hygroscopic |
Boron oxide | |
1303-86-2 | |
Powder | |
99.999% | |
MFCD00011315 | |
Soluble in water. Slightly soluble in methanol. | |
O=BOB=O | |
69.62 | |
CHEBI:30163 | |
Odorless | |
2.46 |
Chemical Identifiers
1303-86-2 | |
69.62 | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
518682 | |
oxo[(oxoboranyl)oxy]borane |
B2O3 | |
MFCD00011315 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
CHEBI:30163 | |
O=BOB=O |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P201-P202-P281-P308+P313-P501c
H360FD
EINECSNumber : 215-125-8
RTECSNumber : ED7900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only