missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron oxide, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1115836

 View more versions of this product

Catalog No. AA1115836



Description

Description

  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Specifications

Specifications

Boron oxide
1303-86-2
B2O3
14,1337
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo(oxoboranyloxy)borane
518682
69.62
99.999%
2.46g/mL (±)0.01
Hygroscopic
White
500g
Fused Lump, Approx. 1.0 in. dia. x 0.75 in. thick
(metals basis)
MFCD00011315
boric acid anhydride
B(=O)OB=O
69.617
CHEBI:30163
Fused Lump
Puratronic™
Odorless
1860°C
~450°C

Chemical Identifiers

1303-86-2
69.617
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
oxo(oxoboranyloxy)borane
B2O3
MFCD00011315
boric acid anhydride
CHEBI:30163
B(=O)OB=O
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : ED7900000

TSCA : TSCA

Documents