Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron oxide, Puratronic™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Boron oxide
CAS 1303-86-2
Name Note Fused Lump, Approx. 1.0 in. dia. x 0.75 in. thick
Purity Grade Puratronic™
Formula Weight 69.62
MDL Number MFCD00011315
Merck Index 14,1337
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1115822

  SDS
alfa aesar™
1115822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1115836

  SDS
alfa aesar™
1115836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 69.62
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.
Name Note Fused Lump, Approx. 1.0 in. dia. x 0.75 in. thick
Odor Odorless
Assay 99.999%
Merck Index 14,1337
TSCA TSCA
CAS 1303-86-2
Melting Point ≈450°C
Assay Percent Notes (metals basis)
Physical Form Fused Lump
RTECS Number ED7900000
Sensitivity Hygroscopic
MDL Number MFCD00011315
Purity Grade Puratronic™
Color White
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Boron oxide
Boiling Point 1860°C
Health Hazard 1 Danger
Density 2.46g/mL (±)0.01
Quantity 100g

  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)

Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.