missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron oxide, (H2O 200ppm), 99.999% (metals basis), Alfa aesar™ Puratronic™

For Use in the Electronics Industry As a Liquid Encapsulent For Czochralski-Type Growth of III-V Semiconductor Single Crystals (E.G., Gaas, Gap

Manufacturer:  Thermo Fisher Scientific 11159QE

 View more versions of this product

Catalog No. AA11159QE



Specifications

Specifications

Boron oxide
1303-86-2
B2O3
White cylinder
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo(oxoboranyloxy)borane
518682
69.62
2.46±0.01g/cm3 at 20°C
White
(H2O 200ppm)
99.999% (metals basis)
MFCD00011315
boric acid anhydride
B(=O)OB=O
69.617
CHEBI:30163
Cylinder, 3.7cm dia., 32g/cylinder
Odorless
1 x 32g

Chemical Identifiers

1303-86-2
69.617
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
oxo(oxoboranyloxy)borane
B2O3
MFCD00011315
boric acid anhydride
CHEBI:30163
B(=O)OB=O
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H360: May damage fertility or the unborn child.

EINECSNumber : 215-125-8

Documents