missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron oxide, 99.98% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8996422

 View more versions of this product

Catalog No. AA8996422



Specifications

Specifications

Boron oxide
99.98% (Metal basis)
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
B(=O)OB=O
69.617
CHEBI:30163
Powder
Odorless
100g
1303-86-2
B2O3
White powder
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo(oxoboranyloxy)borane
518682
69.62
2.46g/cm3
White

Chemical Identifiers

1303-86-2
69.617
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
oxo(oxoboranyloxy)borane
B2O3
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
CHEBI:30163
B(=O)OB=O
Safety and Handling

Safety and Handling

H360

EINECSNumber : 215-125-8

SDS Documents