Learn More
Boron oxide, 98%, -40 mesh, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 206300250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Boron oxide | |
25kg | |
B2O3 | |
MFCD00011315 | |
15,1348 | |
Solubility in water: 36g/L (25°C). Other solubilities: soluble in hot water,soluble in alcohol,glycerol and acids | |
B(=O)OB=O | |
69.61 | |
CHEBI:30163 | |
98% | |
2.46 |
1303-86-2 | |
98% | |
B2O3 | |
01,673; 02,32; 03,33; 09,1349 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
oxo(oxoboranyloxy)borane | |
518682 | |
69.61 | |
Fibre drum | |
2.4600g/mL |
Safety and Handling
GHS H Statement:
May damage fertility. May damage the unborn child.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 215-125-8
RUO â Research Use Only