missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron oxide, 97.5% min., typically 98.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1229036

 View more versions of this product

Catalog No. AA1229036



Specifications

Specifications

Boron oxide
97.5% min., typically 98.5%
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo(oxoboranyloxy)borane
518682
69.62
2.46 ^+ 0.01g/mL
1303-86-2
B2O3
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
B(=O)OB=O
69.617
CHEBI:30163
-60 Mesh
500g

Chemical Identifiers

1303-86-2
69.617
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
oxo(oxoboranyloxy)borane
B2O3
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
CHEBI:30163
B(=O)OB=O
Safety and Handling

Safety and Handling

EINECSNumber : 215-125-8

Documents