Learn More
Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012290A9
Description
It is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from Talc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1860°C | |
∽450°C | |
10 kg | |
B2O3 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
oxo[(oxoboranyl)oxy]borane | |
518682 | |
69.62 | |
2.46 |
Boron oxide | |
1303-86-2 | |
97.5% min., typically 98.5% | |
MFCD00011315 | |
Slightly soluble in water and methanol. | |
O=BOB=O | |
69.62 | |
CHEBI:30163 | |
Hygroscopic |
Chemical Identifiers
1303-86-2 | |
69.62 | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
518682 | |
O=BOB=O |
B2O3 | |
MFCD00011315 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
CHEBI:30163 |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H360FD
EINECSNumber : 215-125-8
RTECSNumber : ED7900000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only