Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron, Oil based standard solution, Specpure™, B 1000μg/g

Chemical Identifiers

Chemical Name or Material Boron Oil based standard solution
Name Note B 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00134034
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1422718

  SDS
alfa aesar™
1422718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note B 1000μg/g
MDL Number MFCD00134034
Purity Grade Specpure™
Chemical Name or Material Boron Oil based standard solution
Physical Form Liquid
Quantity 50g