Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Boron nitride sputtering target
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 24.82
MDL Number MFCD00011317
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41054KS

 
alfa aesar™
41054KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 24.82
Chemical Name or Material Boron nitride sputtering target
TSCA TSCA
Odor Odorless
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011317
Quantity 1 each