missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron nitride, Refractory Brushable Paint, BN 23%

Manufacturer:  Alfa Aesar™ 43279A7

 View more versions of this product

Catalog No. AA43279A7



Specifications

Specifications

Boron nitride
10043-11-5
MFCD00011317
PZNSFCLAULLKQX-UHFFFAOYSA-N
azanylidyneborane
66227
Paste
Odorless
5kg
Refractory Brushable Paint
BN
boron nitride, elbor, boron nitride, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
B#N
24.817
CHEBI:50883
BN 23%
White

Chemical Identifiers

10043-11-5
24.817
PZNSFCLAULLKQX-UHFFFAOYSA-N
66227
azanylidyneborane
BN
MFCD00011317
boron nitride, elbor, boron nitride, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
CHEBI:50883
B#N
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents