missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron nitride, Refractory Brushable Paint, BN 10%

Manufacturer:  Alfa Aesar™ 43278A1

 View more versions of this product

Catalog No. AA43278A1



Specifications

Specifications

Boron nitride
10043-11-5
MFCD00011317
PZNSFCLAULLKQX-UHFFFAOYSA-N
azanylidyneborane
66227
Paste
Odorless
1kg
Refractory Brushable Paint
BN
borazon, boron nitride, denka boron nitride gp, elbor, elboron, geksanit r, hexanite r, kubonit, wurzin
B#N
24.817
CHEBI:50883
BN 10%
White

Chemical Identifiers

10043-11-5
24.817
PZNSFCLAULLKQX-UHFFFAOYSA-N
66227
azanylidyneborane
BN
MFCD00011317
borazon, boron nitride, denka boron nitride gp, elbor, elboron, geksanit r, hexanite r, kubonit, wurzin
CHEBI:50883
B#N
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents