Boron nitride paste, Alfa Aesar™
Has Resistance Against Molten Metals and Salts, High thermal Conductivity and Dielectric Strength
Manufacturer:  Alfa Aesar 4377314
 View more versions of this product
Catalog No. AA4377314
Specifications
Specifications
Boron nitride
BN
White paste
PZNSFCLAULLKQX-UHFFFAOYSA-N
azanylidyneborane
66227
Odorless
25g
10043-11-5
MFCD00011317
borazon
B#N
24.817
CHEBI:50883
White
Chemical Identifiers
10043-11-5
24.817
PZNSFCLAULLKQX-UHFFFAOYSA-N
66227
azanylidyneborane
BN
MFCD00011317
borazon
CHEBI:50883
B#N
Safety and Handling
Safety and Handling
H319-H335: Causes serious eye irritation. May cause respiratory irritation.
Documents