Learn More
Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals
Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.
Supplier: Thermo Scientific Chemicals 045573KG
Description
Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boron nitride | |
Hot Pressed Rod | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
boranylidyneamine | |
66227 | |
24.82 | |
Bag |
10043-11-5 | |
1 Pc. | |
MFCD00011317 | |
Insoluble in water. Boron nitride is insoluble in the acids. Soluble in alkaline molten salts and nitrides, | |
B=N | |
24.82 | |
CHEBI:50883 | |
99.5% |
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
boranylidyneamine |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 | |
B=N |
Safety and Handling
P261-P271-P304+P340-P312-P501c
H335
EINECSNumber : 233-136-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only